
5-(Methoxycarbonyl)-1H-pyrrole-3-boronic acid, pinacol ester, N-BOC protected 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:942070-38-4
:
Formula:C₁₇H₂₆BNO₆
MDL Number:MFCD11855974
:351.202 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together