Lithium 2-[(4-methylpiperazin-1-yl)methyl]benzoate 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:915707-44-7
:
Formula:C₁₃H₁₇N₂O₂·Li
MDL Number:MFCD09064954
:240.227 g/mol
Melting Pt:337…348.5 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together