Order Entry
France
ContactUsLinkComponent
Thieno[2,3-b]pyridine-2-carbohydrazide
Thieno[2,3-b]pyridine-2-carbohydrazide
Catalog # APOSOR111264-1G
Supplier:  Apollo Scientific
CAS Number:  
Thieno[2,3-b]pyridine-2-carbohydrazide
Catalog # APOSOR111264-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.226 g/mol
Storage Temperature:Ambient
Formula:C₈H₇N₃OS
MDL Number:MFCD29088967