Order Entry
France
ContactUsLinkComponent
4-(4-Isopropylphenyl)-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
4-(4-Isopropylphenyl)-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
Catalog # APOSOR111063-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(4-Isopropylphenyl)-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
Catalog # APOSOR111063-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :303.422 g/mol
Storage Temperature:Ambient
CAS Number:1858251-43-0
Formula:C₁₆H₂₁N₃OS
MDL Number:MFCD28148238