
Tropinone oxime hydrochloride 97%
:212079-30-6
:
:C₈H₁₄N₂O·ClH
:MFCD01073570
:190.67 g/mol
:GHS07
:Refrigerator
Frequently Bought Together
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals