
N-(3-Indoleacetyl)-DL-aspartic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:32449-99-3
:
Formula:C₁₄H₁₄N₂O₅
MDL Number:MFCD00050396
:290.271 g/mol
Melting Pt:198 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together