Hesperetin 7-O-β-D-glucuronide ≥95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1237479-09-2
:
Storage Temperature:Ambient
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Pasteur pipettes, disposable
Supplier: Brand
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Cell culture storage bottles, square, PC, Corning®
Supplier: Corning
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection