Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methyl-DL-tryptophan
6-Methyl-DL-tryptophan
6-Methyl-DL-tryptophan
APOSBIB6113-1G
:
Apollo Scientific
:
6-Methyl-DL-tryptophan
APOSBIB6113-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
290 °C
:
GHS07
:
Refrigerator
:
2280-85-5
:
MFCD00022772