UNC-2025 95%
Catalog # ACRO467881000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to off-white Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:476.66 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:1429881-91-3
Formula:C₂₈H₄₀N₆O
MDL Number:MFCD28142874