Order Entry
France
ContactUsLinkComponent
Allergen protein ELISA kits
Allergen protein ELISA kits
Supplier:  3M Food Safety
Certificates
undefined
Allergen protein ELISA kits
Supplier:  3M Food Safety
Certificates

Specifications

  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Target protein:
    Allergen: Bovine casein
  • Detection method:
    Colorimetric (450 nm)
  • Detection limit:
    Protein threshold: 0,5 ppm

Specifications

Related Information

Frequently Bought Together