Order Entry
France
ContactUsLinkComponent
Coumarin 314 97% (dye content), Sigma-Aldrich®
Coumarin 314 97% (dye content), Sigma-Aldrich®
Catalog # 392995-500MG
CAS Number:  
Coumarin 314 97% (dye content), Sigma-Aldrich®
Catalog # 392995-500MG
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    70.0 - 73.7 %
  • Nitrogen:
    6.2 - 6.6 %
  • Wavelength, c = 0.01 g/L; Ethanol:
    433 - 439 nm
  • Extinction Coefficient:
    ≥39000

Specification Test Results

About this item

 :
 :313.35 g/mol
Melting Pt:140...144 °C, 146,22...147,36 °C (lit.)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
EINECS:259-825-1
CAS Number:55804-66-5
Formula:C₁₈H₁₉NO₄
MDL Number:MFCD00051334