Order Entry
France
ContactUsLinkComponent
Acetaldehyde 2,4-dinitrophenylhydrazone analytical standard, for environmental analysis, Sigma-Aldrich®
Irritant
Acetaldehyde 2,4-dinitrophenylhydrazone analytical standard, for environmental analysis, Sigma-Aldrich®
Référence 429554-20MG
CAS Number:  
undefined
Irritant
Acetaldehyde 2,4-dinitrophenylhydrazone analytical standard, for environmental analysis, Sigma-Aldrich®
Référence 429554-20MG
CAS Number:  

Specifications

  • Pk:
    20 mg
  • Seq:
    2305

Specifications

About this item

 :
Melting Pt:165…168 °C (lit.)
 :224.18 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1019-57-4
 :MFCD00458721
Formula:C₈H₈N₄O₄
MDL Number:MFCD00191298