Malonaldehydebis(phenylimine) monohydrochloride 97%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:123071-42-1
:
Formula:C₁₅H₁₄N₂·HCl
MDL Number:MFCD00054634
Melting Pt:218 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
:258.75 g/mol
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (colour):Faint yellow to yellow to beige and faint brown to light brown and faint orange to orange and faint yellow-orange to yellow-orange
- Appearance (form):Powder
- Argentometr. Titration:96.5 - 103.5%
- Purity (HPLC area %):≥96.5%
- Solubility (colour):Yellow to orange
- Solubility (turbidity):Clear
- Solubility (method):2.5%, MeOH
- Infrared spectrum:Conforms to structure
Specification Test Results
Frequently Bought Together