3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester, Sigma-Aldrich®
Catalog # A0223-5MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
Storage Temperature:Freezer
MDL Number:MFCD00467357