Order Entry
France
ContactUsLinkComponent
 
Azure B eosinate, Sigma-Aldrich®
Catalog # 200239-25G
CAS Number:  
Azure B eosinate, Sigma-Aldrich®
Catalog # 200239-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • TLC:
    Consistent with Past Lots
  • Solubility (Color) c = 1mg/ml, Acetic Acid:
    Dark Blue
  • Wavelength c = 0.005 g/l in Methanol:
    635 - 641 nm
  • Extinction Coefficient:
    ≥142400
  • Wavelength:
    521 - 527 nm

Specification Test Results

About this item

 :
 :1186.62 g/mol
Storage Temperature:Ambient
EINECS:263-490-7
CAS Number:62298-42-4
Formula:C₅H₃₈Br₄N₆O₅S₂