Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Brilliant yellow
Brilliant yellow, Sigma-Aldrich®
Brilliant yellow, Sigma-Aldrich®
201375-25G
:
SIGMA ALDRICH MICROSCOPY
:
Brilliant yellow, Sigma-Aldrich®
201375-25G
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
25 g
Seq:
3308
:
:
624.55 g/mol
:
Ambient
:
3051-11-4
:
C₂₆H₂₀N₄O₈S₂
:
MFCD00007489