Order Entry
France
ContactUsLinkComponent
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene 99% for scintillation, Sigma-Aldrich®
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene 99% for scintillation, Sigma-Aldrich®
Catalog # 223999-5G
CAS Number:  
undefined
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene 99% for scintillation, Sigma-Aldrich®
Catalog # 223999-5G
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :430.57 g/mol
Melting Pt:199…201 °C (lit.)
Storage Temperature:Ambient
CAS Number:7128-64-5
Formula:C₂₆H₂₆N₂O₂S
MDL Number:MFCD00005774