Nile blue A ≥75 % (dye content), Sigma-Aldrich®
Catalog # N5632-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:732.84 g/mol
Melting Pt:>300 °C (dec., lit.)
Storage Temperature:Ambient
CAS Number:3625-57-8
Formula:C₄₀H₄₀N₆O₆S
MDL Number:MFCD00064529