Order Entry
France
ContactUsLinkComponent
Haematoxylin solution (Mayer), Sigma-Aldrich®
Irritant
Haematoxylin solution (Mayer), Sigma-Aldrich®
Catalog # MHS1-100ML
CAS Number:  
undefined
Irritant
Haematoxylin solution (Mayer), Sigma-Aldrich®
Catalog # MHS1-100ML
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :302.28 g/mol
 :GHS07
Storage Temperature:Ambient
 :1
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111