Order Entry
France
ContactUsLinkComponent
Acid blue 20, Practical grade, Sigma-Aldrich®
Acid blue 20, Practical grade, Sigma-Aldrich®
Catalog # I2131-25G
CAS Number:  
Acid blue 20, Practical grade, Sigma-Aldrich®
Catalog # I2131-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
CAS Number:8004-99-7
MDL Number:MFCD00131375