Order Entry
France
ContactUsLinkComponent
9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N-hydroxysuccinimide ester chloride, Sigma-Aldrich®
9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N-hydroxysuccinimide ester chloride, Sigma-Aldrich®
Catalog # C6331-1MG
CAS Number:  
undefined
9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N-hydroxysuccinimide ester chloride, Sigma-Aldrich®
Catalog # C6331-1MG
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :567.08 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:158358-57-7
Formula:C₃H₃₅ClN₄O₅
MDL Number:MFCD01310495