
N,N-Diethyl-p-phenylenediamine oxalate salt, Sigma-Aldrich®
Catalog # D5143-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:254.28 g/mol
Melting Pt:145 °C (dec.)
ADR:6.1,III
:GHS07
Storage Temperature:Refrigerator
CAS Number:142439-89-2
Formula:CH₁₆N₂
MDL Number:MFCD00042025