

Fast Blue BB base, Sigma-Aldrich®
Catalog # F0125-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:300.36 g/mol
Melting Pt:98…100 °C
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:120-00-3
Formula:C₁₇H₂₀N₂O₃
MDL Number:MFCD00009091