Order Entry
France
ContactUsLinkComponent
Naphthol AS-D Chloroacetate (Specific Esterase) Kit Select reagents packaged in gelatin capsules., Sigma-Aldrich®
Naphthol AS-D Chloroacetate (Specific Esterase) Kit Select reagents packaged in gelatin capsules., Sigma-Aldrich®
Catalog # 90C2-1KT
CAS Number:  
Naphthol AS-D Chloroacetate (Specific Esterase) Kit Select reagents packaged in gelatin capsules., Sigma-Aldrich®
Catalog # 90C2-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075
  • Environmentally Preferable:

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
UN:3316
 :GHS02
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
 :1