Eflornithine hydrochloride monohydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Eflornithine hydrochloride hydrate, DL-alpha-Difluoromethylornithine Hydrochloride, 2,5-Diamino-2-(difluoromethyl)pentanoic acid hydrochloride hydrate
CAS Number:96020-91-6
:
Formula:C₆H₁₅ClF₂N₂O₃
MDL Number:MFCD00242734
Molecular Weight:236.64 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Water:6 to 9 %
- Titration with HClO4:≥97.5 % (dry substance)
Specification Test Results
Frequently Bought Together