Order Entry
France
Orders LinkContactUsLinkComponent
Ventilation unit for storage cabinets
Ventilation unit for storage cabinets
Supplier:   TRIONYX
Ventilation unit for storage cabinets
Supplier:  TRIONYX
Certificates
Ventilation unit for storage cabinets
Ventilation unit for storage cabinets
3 Options Available Below
Product Details & Documents
Ventilation unit with control switch to adjust the fan air flow.
  • Airflow: 215 m³/hour
  • Dimension: 225×305×195 mm
Delivery information: Includes two outlets to attach to the ventilation of the cabinet and the unit and a flexible pipe to connect the unit to the cabinet.
Product Family Options
Product Information
  • Weight
    Availability
    Price
  • 132-4218
    Ventilation with steel fan, one-meter hose, two clamping collars and 100 mm Ø sleeve
    Ventilation with steel fan, one-meter hose, two clamping collars and 100 mm Ø sleeve
    9 kg
    Discontinued
  • 132-4219
    Flexible pipe, Ø = 100 mm
    Flexible pipe, Ø = 100 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 63,86 €
    63,86 €
    1 * 1 m
       63,86 €
  • TRIOCDS100
    Clamp collar, Ø = 100 mm
    Clamp collar, Ø = 100 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 7,21 €
    7,21 €
    1 * 1 Pce
       7,21 €
  • TRIOCDFCORG
    Active coal filter for corrosive and organic vapours
    Active coal filter for corrosive and organic vapours
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 210,12 €
    210,12 €
    1 * 1 Pce
       210,12 €