Analytical standard, Doxycycline hyclate, reference material, Fluka™
Catalog # HONCR1748-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:24390-14-5
- Description:Doxycycline hyclate
- Description:Doxycycline hyclate
- Form:Neat