Tellurium ICP & ICP-MS standard, (CRMs), Fluka™
Catalog # HONCE1357-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:100 mg/L
- Matrice:2% HNO₃
- Type de conditionnement:Plastic bottle
- Cdt:100 mL
- Élement:Tellure
- Symbole:Te
Specifications
About this item
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).