Thorium ICP Standard, (CRMs), Fluka™
HONCE1062-100ML
:
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Thorium
- Symbol:Th
Certified Reference Material (CRM)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).