


Toluene, dried (max. 50 ppm H₂O), Burdick & Jackson™
Catalog # HONC65520-185L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Stainless steel drum
- Pk:1
- Seq:5491
- Pk Info:185 l
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- APHA:Max. 10
- Density (D 20/20):0.865 - 0.869
- Refractive index (n 20/D):1.496 - 1.498
- Water (Karl Fischer):Max. 50 ppm
Specification Test Results
About this item
:
:92.14 g/mol
Boiling Pt:110.6 °C
Melting Pt:-95 °C
Melting Pt:–95 °C
Density:0.867 g/cm3 (20 °C)
Storage Temperature:Ambient
UN:1294
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512