

Pyridine, EMPARTA® ACS analytical reagent
Supplier: Merck
Certificates
About this item
CAS Number:110-86-1
:
Formula:C₅H₅N
MDL Number:MFCD00011732
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–42 °C
Density:0.982 g/cm3 (20 °C)
Flash Pt:17 °C
EINECS:203-809-9
ADR:3,II
:79.1 g/mol
UN:1282
:GHS02
:GHS07
Frequently Bought Together