

4-Nitro-3-picoline-N-oxide 98%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1074-98-2
:
Formula:C₆H₆N₂O₃
MDL Number:MFCD00014626
:154.13 g/mol
Melting Pt:137.5 °C
Storage Temperature:Room Temperature
EINECS:214-050-8
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS07
:GHS08
Specification Test Results
- Appearance (Colour):Yellow
- Appearance (form):Powder or Crystals or Chunk(s)
- Purity (HPLC Area %):≥97.5%
- Carbon content:45.6 - 47.9%
- Nitrogen content:17.7 - 18.6%
- Infrared spectrum:Conforms to structure
Specification Test Results
Frequently Bought Together