
Methylene blue for microscopy
VWRS3470.1000
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Proportion A635/A663:0.54 - 0.62
:
:319.86 g/mol
:12,06137
:180 °C
:1.757 g/cm³ (22 °C)
:200-515-2
:GHS07
:Ambient
:61-73-4
:MFCD00012111
:C₁₆H₁₈ClN₃S
:MFCD00150008