Benzyl phenyl sulphide for synthesis, Sigma-Aldrich®
Catalog # 8.41566.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Melting Point:40 - 44 °C
- Molar Mass:200.29 g/mol
- Hill Formula:C13H12S
Specification Test Results
About this item
:
:200.3
Boiling Pt:197 °C (1013 hPa)
Melting Pt:40…44 °C
:M-Clarity™ quality level = MQ200
EINECS:212-612-7
CAS Number:831-91-4
Formula:C₁₃H₁₂S
MDL Number:MFCD00003066