
Tetradecyltrimethylammonium bromide
Catalog # PRLS85216.260
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:336.394 g/mol
Merck Index:12,06419
Melting Pt:245…250 °C
Storage Temperature:Ambient
EINECS:214-291-9
UN:3261
ADR:8,II
:GHS05
CAS Number:1119-97-7
:8044-71-1
Formula:C₁₇H₃₈BrN
MDL Number:MFCD00011770