Benzyl butyrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:103-37-7
:
Formula:C₁₁H₁₄O₂
MDL Number:MFCD00027133
:178.23 g/mol
Boiling Pt:240 °C (1013 hPa)
Density:1.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-105-1
Specification Test Results
- Solubility:(20 °C) low
- Molar Mass:178.23 g/mol
- Boiling Point:235 - 242 °C (1013 hPa)
- Vapor Pressure:11.97 hPa (109 °C)
- Flash Point:107 °C
- Density:1.01 g/cm3
Specification Test Results
Frequently Bought Together