Order Entry
France
ContactUsLinkComponent
4,4'-Bis(2-methoxystyryl)biphenyl ≥90.0% (by HPLC)
4,4'-Bis(2-methoxystyryl)biphenyl ≥90.0% (by HPLC)
Catalog # TCIAB2730-25G
Supplier:  TCI
CAS Number:  
4,4'-Bis(2-methoxystyryl)biphenyl ≥90.0% (by HPLC)
Catalog # TCIAB2730-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystaline
  • Purity (HPLC):
    min. 90.0 area%
  • Melting point:
    217.0 to 221.0 deg-C

Specification Test Results

About this item

Synonyms: 4,4-Bis(2-methoxystyryl)biphenyl, 4,4'-Bis[2-(2-methoxyphenyl)vinyl]biphenyl
 :
 :418.54 g/mol
EINECS:254-935-6
CAS Number:40470-68-6
Formula:C₃₀H₂₆O₂
MDL Number:MFCD23160777