Malononitrile for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-77-3
:
Formula:C₃H₂N₂
MDL Number:MFCD00001883
:66.06 g/mol
Merck Index:13,05733
Boiling Pt:220 °C (1013 hPa)
Melting Pt:30…32 °C
Density:1.191 g/cm³ (20 °C)
EINECS:203-703-2
UN:2647
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:112 °C
:GHS06
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:133 g/l (20 °C)
- Melting Point:32 - 34 °C
- Molar Mass:66.06 g/mol
- Boiling Point:220 °C (1013 hPa)
- Vapor Pressure:1 hPa (50 °C)
- Flash Point:112 °C
- Density:1.05 g/cm3 (20 °C)
- Ignition Point:365 °C
Specification Test Results
Frequently Bought Together