Order Entry
France
ContactUsLinkComponent
1-Cetylpyridinium chloride monohydrate 96.0-101.0%
undefined
1-Cetylpyridinium chloride monohydrate 96.0-101.0%
  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    4.5 to 5.5 %
  • Titration:
    96.0 to 101.0 %
Frequently Bought Together