Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
Frequently Bought Together