Calcium acetate xhydrate, Supelco®
Catalog # 1.09325.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Solubility:400 g/l (0 °C)
- Molar Mass:158.17 g/mol (anhydrous)
- Bulk Density:400 kg/m3
- Density:1.5 g/cm3 (20 °C)
- pH:7.2 - 7.8 (50 g/l, H2O, 20 °C)
- Ignition Point:680 - 730 °C
Specification Test Results
About this item
:
:176.18 g/mol
Density:1.5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:200-540-9
CAS Number:114460-21-8
:MFCD00012448
Formula:(H₃CCOO)₂Ca·nH₂O
MDL Number:MFCD00150019