Order Entry
France
Orders LinkContactUsLinkComponent
Magnesium sulphate 0.1 mol/l in aqueous solution

Magnesium sulphate 0.1 mol/l in aqueous solution

Plastic bottle, 500 mL
Catalog # PRLSMC312677.0500
Supplier:   PROLABO CMP
Magnesium sulphate 0.1 mol/l in aqueous solution
Catalog # PRLSMC312677.0500
Supplier:  PROLABO CMP
CAS Number:  7487-88-9
Magnesium sulphate 0.1 mol/l in aqueous solution
Magnesium sulphate 0.1 mol/l in aqueous solution

Plastic bottle, 500 mL

CAS Number:  7487-88-9
Price
37,90 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  MgSO4, Magnesium sulphate anhydrous
  • CAS Number:7487-88-9
  • Formula:MgSO₄
  • MDL Number:MFCD00011110
  • Molecular Weight:120.37 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-298-2

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.