Sodium hydroxide 0.01 M NaOH (0.01N) volumetric solution, Fluka™
Catalog # 35262-10L-VP
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:VOLPAK container
- Pk:10 L
- Seq:5214
Specifications
Specification Test Results
- Factor:0.995 - 1.005
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548