Potassium hydroxide 1 mol/l (1 N) in aqueous solution
PRLSMC307195.0500
:
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
: Potash caustic, Caustic potash
:
:56.11 g/mol
:Ambient
:215-181-3
:1814
:8,III
:GHS05
:1310-58-3
:KOH
:MFCD00003553