Order Entry
France
ContactUsLinkComponent
Potassium permanganate 0.1 mol/l (0.5 N) in aqueous solution
Environment
Potassium permanganate 0.1 mol/l (0.5 N) in aqueous solution
Catalog # FFCHFF7315000
Supplier:  FF CHEMICALS
CAS Number:  
Environment
Potassium permanganate 0.1 mol/l (0.5 N) in aqueous solution
Catalog # FFCHFF7315000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364