Order Entry
France
ContactUsLinkComponent
Organic reference standard, 4-Nitrophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 4-Nitrophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15590400ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 4-Nitrophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15590400ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    100-02-7
  • Description:
    4-Nitrophenol 100 µg/ml in methanol
  • Formula:
    C₆H₅NO₃
  • Description:
    4-Nitrophenol

Specifications

About this item