Molybdenum standard solution, 1000 mg/L Mo in water (from (NH₄)₆Mo₇O₂₄) Certipur® standard for ICP, Supelco®
Référence 1.70334.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Molybdenum
- Symbol:Mo
Specifications
About this item
Certifications: Traceable to SRM from NIST.