Lithium standard solution, 1000 mg/L Li in dil. nitric acid (from LiNO₃) Certipur® standard for ICP, Supelco®
Référence 1.70329.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Lithium
- Symbol:Li
Specifications
About this item
Certifications: Traceable to SRM from NIST.