DL-Lysine monohydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70-53-1
:
Formula:C₆H₁₅ClN₂O₂
MDL Number:MFCD00064563
Melting Pt:267 °C
:182.65 g/mol
Storage Temperature:Ambient
EINECS:200-739-0
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Specific optical rotation:-1° to +1° (20°C, 589 nm) (c=1, water)
- Titration with HClO4:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together