Order Entry
France
ContactUsLinkComponent
Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Catalog # 31963.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Catalog # 31963.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

Synonyms: Fehling A solution
 :
 :159.61 g/mol
Boiling Pt:>100 °C (1013 hPa)
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3082
ADR:9,III
 :GHS09
CAS Number:7758-98-7
Formula:CuSO₄
MDL Number:MFCD00010981